Gene Transcripts

Gene Symbol
Adcy6
Entrez Gene ID
25289
Gene Name
adenylate cyclase 6
Gene Aliases
ACVI, ADCYB
Chromosome Location
Chr. 7: 140270692 - 140291620 on Build Rnor_6.0
UniGene ID
Rn.3313
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270785.1 NP_001257714.1 2 863
GenBank mRNA L01115.1 - 2 1006
M96160.1 - 2 863

View Full Product Details