Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016995.2 | NP_058691.2 | 4 | 739 | |
XM_006249705.2 | XP_006249767.1 | 5 | 795 | ||
XM_008769423.1 | XP_008767645.1 | 5 | 612 | ||
GenBank mRNA | BC088152.1 | - | 4 | 739 | |
FQ209636.1 | - | 4 | 750 | ||
FQ218474.1 | - | 4 | 720 | ||
Z50052.1 | - | 4 | 718 |