Gene Transcripts

Gene Symbol
MKRN2
Entrez Gene ID
23609
Gene Name
makorin ring finger protein 2
Gene Aliases
HSPC070, RNF62
Chromosome Location
Chr. 3: 12557014 - 12583713 on Build GRCh38
UniGene ID
Hs.591666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271707.1 NP_001258636.1 7 2394
NM_014160.4 NP_054879.3 8 2523
GenBank mRNA AF302084.1 - 8 2437
AL834512.1 - 5 2045
BC001799.2 - 6 2230
BC015715.1 - 8 2465 3916199

View Full Product Details