Gene Transcripts

Gene Symbol
B3GNT8
Entrez Gene ID
374907
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8
Gene Aliases
B3GALT7, BGALT15, beta3Gn-T8
Chromosome Location
Chr. 19: 41425359 - 41429024 on Build GRCh38
UniGene ID
Hs.441681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198540.2 NP_940942.1 3 1763
XM_011526934.2 XP_011525236.1 2 3082
GenBank mRNA AY277592.2 - 3 1763

View Full Product Details