Gene Transcripts

Gene Symbol
AGMO
Entrez Gene ID
392636
Gene Name
alkylglycerol monooxygenase
Gene Aliases
TMEM195
Chromosome Location
Chr. 7: 15117223 - 15562015 on Build GRCh38
UniGene ID
Hs.670634
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004320.1 NP_001004320.1 13 1830
GenBank mRNA AK131284.1 - 13 1830

View Full Product Details