Gene Transcripts

Gene Symbol
LINC00467
Entrez Gene ID
84791
Gene Name
long intergenic non-protein coding RNA 467
Gene Aliases
C1orf97
Chromosome Location
Chr. 1: 211382755 - 211432535 on Build GRCh38
UniGene ID
Hs.523932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026761.2 3 388
GenBank mRNA AK130857.1 - 3 355
AK130858.1 - 3 340
BC005997.1 - 3 324
BP197632.1 - 3 389
BX382054.2 - 3 323

View Full Product Details