Gene Transcripts

Gene Symbol
Armc5
Entrez Gene ID
361653
Gene Name
armadillo repeat containing 5
Gene Aliases
Chromosome Location
Chr. 1: 199655647 - 199662414 on Build Rnor_6.0
UniGene ID
Rn.15020
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009455.1 NP_001009455.1 4 1949
GenBank mRNA BC087086.1 - 4 1949

View Full Product Details