Gene Transcripts

Gene Symbol
Cxadr
Entrez Gene ID
89843
Gene Name
coxsackie virus and adenovirus receptor
Gene Aliases
Car1
Chromosome Location
Chr. 11: 16826298 - 16873564 on Build Rnor_6.0
UniGene ID
Rn.113837
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053570.1 NP_446022.1 8 1275
GenBank mRNA BC088313.1 - 8 1275

View Full Product Details