Gene Transcripts

Gene Symbol
Aga
Entrez Gene ID
11593
Gene Name
aspartylglucosaminidase
Gene Aliases
AW060726
Chromosome Location
Chr. 8: 53511687 - 53523422 on Build GRCm38
UniGene ID
Mm.334535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005847.2 NP_001005847.1 1 117
NM_001205054.1 NP_001191983.1 1 117
GenBank mRNA BC048230.1 - 1 21
BC132227.1 - 1 92 40130504
BC132229.1 - 1 92 40130506
BC145297.1 - 1 92 9053851
CD541877.1 - 1 93
S81393.1 - 1 34

View Full Product Details