Gene Transcripts

Gene Symbol
C7orf43
Entrez Gene ID
55262
Gene Name
chromosome 7 open reading frame 43
Gene Aliases
Chromosome Location
Chr. 7: 100154420 - 100158721 on Build GRCh38
UniGene ID
Hs.533139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303470.1 NP_001290399.1 9, 10 1125
NM_018275.4 NP_060745.3 9, 10 1625
GenBank mRNA AK054872.1 - 9, 10 1121
AK302342.1 - 7, 8 973
AL136555.1 - 2, 3 324
BC002749.2 - 9, 10 1025
BC015722.2 - 9, 10 1583 3920412

View Full Product Details