Gene Transcripts

Gene Symbol
CATSPERG
Entrez Gene ID
57828
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
C19orf15
Chromosome Location
Chr. 19: 38335563 - 38370949 on Build GRCh38
UniGene ID
Hs.324335
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021185.4 NP_067008.3 13 1483
XM_005259114.4 XP_005259171.1 13 1579
XM_005259115.4 XP_005259172.1 13 1502
XM_005259116.2 XP_005259173.1 11 1303
XM_006723308.3 XP_006723371.1 13 2099
XM_006723310.2 XP_006723373.1 13 1366
XM_006723311.2 XP_006723374.1 13 1504
XM_011527174.2 XP_011525476.1 13 1493
XM_011527175.2 XP_011525477.1 13 1399
XM_011527176.2 XP_011525478.1 13 1402
XM_011527177.2 XP_011525479.1 13 1363
XM_011527178.2 XP_011525480.1 14 1845
XM_011527179.1 XP_011525481.1 13 1483
XM_011527182.2 XP_011525484.1 12 1227
XM_017027053.1 XP_016882542.1 12 1363
XM_017027054.1 XP_016882543.1 11 1096
GenBank mRNA AK128220.1 - 13 1266
AK310460.1 - 9 813
AL133023.1 - 13 2099
BC094785.1 - 13 1403
BC109035.1 - 9 882 40028284

View Full Product Details