Gene Transcripts
- Gene Symbol
- CAMK4
- Entrez Gene ID
- 814
- Gene Name
- calcium/calmodulin dependent protein kinase IV
- Gene Aliases
- CaMK IV, CaMK-GR, caMK
- Chromosome Location
- Chr. 5: 111223653 - 111498503 on Build GRCh38
- UniGene ID
- Hs.438801
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001323374.1 |
NP_001310303.1
|
10, 11 |
929 |
|
NM_001323375.1 |
NP_001310304.1
|
10, 11 |
961 |
|
NM_001323376.1 |
NP_001310305.1
|
7, 8 |
931 |
|
NM_001323377.1 |
NP_001310306.1
|
8, 9 |
689 |
|
NM_001744.5 |
NP_001735.1
|
9, 10 |
1077 |
|
GenBank mRNA |
AK311662.1 |
- |
7, 8 |
786 |
|
AK312840.1 |
- |
9, 10 |
932 |
|
BC016695.1 |
- |
9, 10 |
921 |
|
BC025687.1 |
- |
9, 10 |
919 |
5205478 |
D30742.1 |
- |
9, 10 |
907 |
|
L17000.1 |
- |
9, 10 |
871 |
|
L24959.1 |
- |
10, 11 |
929 |
|
View Full Product Details