Gene Transcripts
- Gene Symbol
- C17orf75
- Entrez Gene ID
- 64149
- Gene Name
- chromosome 17 open reading frame 75
- Gene Aliases
- NJMU-R1, SRI2
- Chromosome Location
- Chr. 17: 32328441 - 32350047 on Build GRCh38
- UniGene ID
- Hs.655257
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022344.3 |
NP_071739.2
|
8, 9 |
949 |
|
XM_005258022.3 |
XP_005258079.1
|
9, 10 |
967 |
|
XM_017024940.1 |
XP_016880429.1
|
8, 9 |
818 |
|
XM_017024941.1 |
XP_016880430.1
|
8, 9 |
816 |
|
GenBank mRNA |
AB062437.1 |
- |
2, 3 |
171 |
|
AF052093.1 |
- |
8, 9 |
876 |
|
AF305686.1 |
- |
8, 9 |
905 |
|
BC026017.2 |
- |
8, 9 |
895 |
3352761 |
BC035715.1 |
- |
8, 9 |
903 |
5588164 |
BC047325.1 |
- |
8, 9 |
889 |
5493477 |
BC054035.1 |
- |
8, 9 |
883 |
6143106 |
View Full Product Details