Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016474.4 | NP_057558.3 | 5 | 518 | |
NR_135523.1 | 5 | 518 | |||
XM_017006554.1 | XP_016862043.1 | 5 | 581 | ||
XM_017006555.1 | XP_016862044.1 | 5 | 579 | ||
GenBank mRNA | AF151046.1 | - | 5 | 480 | |
AK309525.1 | - | 5 | 491 | ||
BC013999.2 | - | 5 | 494 | 3936914 | |
BC033897.1 | - | 5 | 505 | 4812977 | |
BP195134.1 | - | 5 | 518 | ||
CD580179.2 | - | 5 | 516 |