Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052813.4 | NP_434700.2 | 7 | 1168 | |
NM_052814.3 | NP_434701.1 | 7 | 1168 | ||
GenBank mRNA | AF311287.1 | - | 7 | 1129 | |
AK024001.1 | - | 7 | 1148 | ||
AK127481.1 | - | 2 | 930 | ||
AK292081.1 | - | 7 | 1120 | ||
AK295647.1 | - | 7 | 1007 | ||
BC008877.2 | - | 7 | 1143 | 4139529 |