Gene Transcripts

Gene Symbol
Bora
Entrez Gene ID
306102
Gene Name
bora, aurora kinase A activator
Gene Aliases
RGD1309522
Chromosome Location
Chr. 15: 83442177 - 83464315 on Build Rnor_6.0
UniGene ID
Rn.229814
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013997.1 NP_001014019.1 10 1156
XM_008770868.1 XP_008769090.1 10 1514
GenBank mRNA BC088204.1 - 10 1156

View Full Product Details