Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040032.1 | NP_001035121.1 | 2 | 189 | |
XM_011543816.2 | XP_011542118.1 | 2 | 290 | ||
GenBank mRNA | AK131443.1 | - | 2 | 173 | |
BC065837.1 | - | 2 | 189 | ||
BC132852.1 | - | 2 | 189 | 40146874 | |
BC132854.1 | - | 2 | 189 | 40146876 |