Gene Transcripts

Gene Symbol
ANKUB1
Entrez Gene ID
389161
Gene Name
ankyrin repeat and ubiquitin domain containing 1
Gene Aliases
C3orf16
Chromosome Location
Chr. 3: 149760648 - 149792823 on Build GRCh38
UniGene ID
Hs.665988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144960.2 NP_001138432.1 2 625
NM_001315505.1 NP_001302434.1 2 597
NM_001315506.1 NP_001302435.1 2 625
XM_011512798.1 XP_011511100.1 2 122
XM_011512799.1 XP_011511101.1 2 356
XM_011512802.1 XP_011511104.1 2 356
GenBank mRNA AK098078.1 - 1 81
AK304359.1 - 2 356
BC171805.1 - 2 597

View Full Product Details