Gene Transcripts

Gene Symbol
C4orf48
Entrez Gene ID
401115
Gene Name
chromosome 4 open reading frame 48
Gene Aliases
CHR4_55
Chromosome Location
Chr. 4: 2035610 - 2043970 on Build GRCh38
UniGene ID
Hs.446474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001141936.2 NP_001135408.2 4 469
NM_001168243.1 NP_001161715.1 4 419
XM_011513469.2 XP_011511771.1 5 1405
XM_017008190.1 XP_016863679.1 5 478
GenBank mRNA BC062790.1 - 4 405
BQ129383.1 - 4 375

View Full Product Details