Gene Transcripts

Gene Symbol
ANKRD30B
Entrez Gene ID
374860
Gene Name
ankyrin repeat domain 30B
Gene Aliases
NY-BR-1.1
Chromosome Location
Chr. 18: 14748240 - 14855465 on Build GRCh38
UniGene ID
Hs.567889
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145029.1 NP_001138501.1 7 1194
XM_006722325.3 XP_006722388.1 7 1022
XM_011525662.2 XP_011523964.1 7 1022
XM_011525664.2 XP_011523966.1 7 1022
XM_011525665.2 XP_011523967.1 7 1099
XM_011525666.2 XP_011523968.1 7 1099
GenBank mRNA AF269088.1 - 7 1195
BC028407.1 - 1102

View Full Product Details