Gene Transcripts

Gene Symbol
BOLA2-SMG1P6
Entrez Gene ID
107282092
Gene Name
BOLA2-SMG1P6 readthrough
Gene Aliases
Chromosome Location
Chr. 16: 29442905 - 29454964 on Build GRCh38
UniGene ID
Hs.444600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320622.1 NP_001307551.1 1 593
NM_001320623.1 NP_001307552.1 1 593
NM_001320624.1 NP_001307553.1 1 593
NM_001320625.1 NP_001307554.1 1 593
NM_001320627.1 NP_001307556.1 1 593
NR_135316.1 1 593
GenBank mRNA AF271775.1 - 1 189
BC130401.1 - 1 280 40146432
BC130403.1 - 1 280 40146434

Gene Symbol
BOLA2
Entrez Gene ID
552900
Gene Name
bolA family member 2
Gene Aliases
BOLA2A, BOLA2B, My016
Chromosome Location
Chr. 16: 29453588 - 29454964 on Build GRCh38
UniGene ID
Hs.444600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031827.2 NP_001026997.1 1 593
NM_001320579.1 NP_001307508.1 1 593
GenBank mRNA BC022832.1 - 1 593
BC144688.1 - 1 280 9053222

Gene Symbol
BOLA2B
Entrez Gene ID
654483
Gene Name
bolA family member 2B
Gene Aliases
BOLA2
Chromosome Location
Chr. 16: 30192930 - 30194306 on Build GRCh38
UniGene ID
Hs.444600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039182.2 NP_001034271.1 1 593
NM_001320607.1 NP_001307536.1 1 593
GenBank mRNA BC137532.1 - 1 280
BC137534.1 - 1 280 9021534

View Full Product Details