Gene Transcripts
- Gene Symbol
- C19orf60
- Entrez Gene ID
- 55049
- Gene Name
- chromosome 19 open reading frame 60
- Gene Aliases
- Chromosome Location
- Chr. 19: 18588685 - 18592337 on Build GRCh38
- UniGene ID
- Hs.655317
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054670.1 |
- |
4 |
1351 |
|
AK091925.1 |
- |
3 |
1655 |
|
AK094710.1 |
- |
1 |
1846 |
|
View Full Product Details