Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001663.3 | NP_001654.1 | 2 | 779 | |
GenBank mRNA | AF047432.1 | - | 1 | 225 | |
AF493885.1 | - | 1 | 158 | ||
AK313790.1 | - | 2 | 698 | ||
BC002952.1 | - | 2 | 682 | ||
BC008918.2 | - | 2 | 694 | 3626100 | |
BT019868.1 | - | 1 | 158 | ||
CR541939.1 | - | 1 | 158 | ||
CR541964.1 | - | 1 | 158 | ||
M57763.1 | - | 2 | 675 |