Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136039.2 | NP_001129511.1 | 2 | 354 | |
NM_001142355.1 | NP_001135827.1 | 2 | 303 | ||
NM_001142356.1 | NP_001135828.1 | 1 | 289 | ||
NM_021824.3 | NP_068596.2 | 2 | 506 | ||
XM_005246742.3 | XP_005246799.1 | 2 | 569 | ||
XM_011511582.1 | XP_011509884.1 | 2 | 490 | ||
XM_011511583.1 | XP_011509885.1 | 2 | 1234 | ||
XM_011511584.1 | XP_011509886.1 | 2 | 1052 | ||
XM_011511585.1 | XP_011509887.1 | 2 | 1001 | ||
XM_017004630.1 | XP_016860119.1 | 2 | 751 | ||
GenBank mRNA | AB038949.1 | - | 2 | 467 | |
AF060513.1 | - | 2 | 263 | ||
AF182416.1 | - | 2 | 425 | ||
AF283538.1 | - | 2 | 309 | ||
AK023378.1 | - | 2 | 452 | ||
AK314709.1 | - | 2 | 308 | ||
AL834430.1 | - | 1 | 129 | ||
AY251943.1 | - | 1 | 292 | ||
AY251944.1 | - | 1 | 292 | ||
BC007654.2 | - | 1 | 290 | 3346099 | |
BG724012.1 | - | 2 | 562 | ||
BP194872.1 | - | 2 | 303 |