Gene Transcripts

Gene Symbol
TEPSIN
Entrez Gene ID
146705
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Gene Aliases
C17orf56, ENTHD2
Chromosome Location
Chr. 17: 81228277 - 81239092 on Build GRCh38
UniGene ID
Hs.631761
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144679.2 NP_653280.1 3, 4 279
XM_005257066.2 XP_005257123.1 4, 5 873
XM_005257067.2 XP_005257124.1 3, 4 279
XM_006721709.3 XP_006721772.1 3, 4 286
XM_006721712.2 XP_006721775.1 3, 4 717
XM_011524355.1 XP_011522657.1 4, 5 879
XM_011524356.1 XP_011522658.1 4, 5 879
XM_011524357.1 XP_011522659.1 3, 4 907
XM_011524358.2 XP_011522660.1 4, 5 808
XM_017024202.1 XP_016879691.1 4, 5 1120
XM_017024203.1 XP_016879692.1 5, 6 767
XM_017024204.1 XP_016879693.1 4, 5 1320
GenBank mRNA AK056090.1 - 3, 4 243
AK127221.1 - 1, 2 1432
AK128728.1 - 4, 5 1592
BC064483.1 - 3, 4 240 5430837
DB122720.1 - 3, 4 279

View Full Product Details