Gene Transcripts

Gene Symbol
C2orf57
Entrez Gene ID
165100
Gene Name
chromosome 2 open reading frame 57
Gene Aliases
Chromosome Location
Chr. 2: 231592864 - 231594283 on Build GRCh38
UniGene ID
Hs.98104
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152614.2 NP_689827.2 1 275
GenBank mRNA BC034405.1 - 1 238
BC063389.1 - 1 238 5170341

View Full Product Details