Gene Transcripts

Gene Symbol
BMT2
Entrez Gene ID
154743
Gene Name
base methyltransferase of 25S rRNA 2 homolog
Gene Aliases
C7orf60
Chromosome Location
Chr. 7: 112819147 - 112939877 on Build GRCh38
UniGene ID
Hs.489734
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152556.2 NP_689769.2 5 1608
GenBank mRNA AK056380.1 - 5 1607
AL834437.1 - 5 1403

View Full Product Details