Gene Transcripts

Gene Symbol
TMEM52B
Entrez Gene ID
120939
Gene Name
transmembrane protein 52B
Gene Aliases
C12orf59
Chromosome Location
Chr. 12: 10170599 - 10191804 on Build GRCh38
UniGene ID
Hs.226422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079815.1 NP_001073283.1 6 764
NM_153022.3 NP_694567.1 4 990
XM_006719029.2 XP_006719092.1 5 1034
GenBank mRNA AK055728.1 - 4 916
AY358845.1 - 4 947
BC098571.1 - 5 474 30921470
BC131523.1 - 5 478 40032531

View Full Product Details