Gene Transcripts

Gene Symbol
EVX1
Entrez Gene ID
2128
Gene Name
even-skipped homeobox 1
Gene Aliases
EVX-1
Chromosome Location
Chr. 7: 27242545 - 27247819 on Build GRCh38
UniGene ID
Hs.369879
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304519.1 NP_001291448.1 1 562
NM_001304520.1 NP_001291449.1 1 562
NM_001989.4 NP_001980.1 1 562
GenBank mRNA AK299188.1 - 1 562
AK314155.1 - 1 407
X60655.1 - 1 304

View Full Product Details