Gene Transcripts

Gene Symbol
Camta1
Entrez Gene ID
100072
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
1810059M14Rik, 2310058O09Rik, AI316882, AW987475, mKIAA0833
Chromosome Location
Chr. 4: 151059523 - 151861768 on Build GRCm38
UniGene ID
Mm.318846
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195565.1 NP_001182494.1 4 1223
GenBank mRNA AK034038.1 - 4 1160
AK040304.1 - 1 678

View Full Product Details