Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028607.1 | NP_082883.1 | 3, 4 | 317 | |
GenBank mRNA | AK010745.1 | - | 3, 4 | 314 | |
AK145313.1 | - | 3, 4 | 318 | ||
BC048500.1 | - | 2, 3 | 197 | ||
BC065806.1 | - | 3, 4 | 310 | 1547643 |