Gene Transcripts

Gene Symbol
Akap4
Entrez Gene ID
11643
Gene Name
A kinase (PRKA) anchor protein 4
Gene Aliases
AKAP-4, AKAP82, Fsc1, PRKA4, mAKAP82, p82
Chromosome Location
Chr. X: 7067515 - 7078609 on Build GRCm38
UniGene ID
Mm.1498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042542.2 NP_001036007.1 2, 3 238
NM_009651.4 NP_033781.2 2, 3 234
GenBank mRNA BC130271.1 - 2, 3 166
CJ047282.1 - 2, 3 236
U07423.1 - 2, 3 207
U10341.1 - 2, 3 239

View Full Product Details