Gene Transcripts

Gene Symbol
Ebi3
Entrez Gene ID
50498
Gene Name
Epstein-Barr virus induced gene 3
Gene Aliases
EBI-3, IL-27
Chromosome Location
Chr. 17: 55952623 - 55957022 on Build GRCm38
UniGene ID
Mm.256798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015766.2 NP_056581.1 5 938
GenBank mRNA AF013114.1 - 5 921
AK155023.1 - 5 901
AK155903.1 - 5 793
BC008209.1 - 5 886 3154957

View Full Product Details