Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025600.1 | NP_001020771.1 | 9 | 1837 | |
NM_018770.3 | NP_061240.3 | 10 | 1921 | ||
NM_207675.2 | NP_997558.2 | 11 | 1954 | ||
NM_207676.2 | NP_997559.1 | 10 | 1870 | ||
GenBank mRNA | AB052293.1 | - | 10 | 1921 | |
AF061260.1 | - | 9 | 1758 | ||
AF434663.1 | - | 10 | 2016 | ||
AK013911.1 | - | 11 | 2049 | ||
AK164133.1 | - | 1 | 153 | ||
BC057125.1 | - | 10 | 1858 | 6414884 | |
BC094661.1 | - | 7 | 1440 | ||
BC095986.1 | - | 10 | 1913 | 5372277 |