Gene Transcripts

Gene Symbol
Cbln3
Entrez Gene ID
56410
Gene Name
cerebellin 3 precursor protein
Gene Aliases
Chromosome Location
Chr. 14: 55878920 - 55884256 on Build GRCm38
UniGene ID
Mm.97163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019820.3 NP_062794.1 2, 3 473
GenBank mRNA AF218379.1 - 2, 3 449
AK081085.1 - 2, 3 449
BC065108.1 - 2, 3 473 30536445

View Full Product Details