Gene Transcripts
- Gene Symbol
- Aurkc
- Entrez Gene ID
- 20871
- Gene Name
- aurora kinase C
- Gene Aliases
- AIE1, AIK3, ARK-3, IAK3, Stk13
- Chromosome Location
- Chr. 7: 6989892 - 7003364 on Build GRCm38
- UniGene ID
- Mm.261572
- Species
- Mus musculus
- Gene Symbol
- Gm7789
- Entrez Gene ID
- 665798
- Gene Name
- predicted pseudogene 7789
- Gene Aliases
- EG665798
- Chromosome Location
- Chr. 7: 10988428 - 10996013 on Build GRCm38
- UniGene ID
- Mm.458907
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA545317.1 |
- |
2, 3 |
265 |
|
View Full Product Details