Gene Transcripts

Gene Symbol
Asic5
Entrez Gene ID
58170
Gene Name
acid-sensing (proton-gated) ion channel family member 5
Gene Aliases
Accn5, Blinac, Inac
Chromosome Location
Chr. 3: 81982293 - 82021233 on Build GRCm38
UniGene ID
Mm.88839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021370.2 NP_067345.1 8 1230
GenBank mRNA AK038321.1 - 7 1282
Y19035.1 - 8 1229

View Full Product Details