Gene Transcripts
- Gene Symbol
- C8B
- Entrez Gene ID
- 732
- Gene Name
- complement component 8, beta polypeptide
- Gene Aliases
- C82
- Chromosome Location
- Chr. 1: 56929210 - 56974383 on Build GRCh38
- UniGene ID
- Hs.391835
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000066.3 |
NP_000057.2
|
7, 8 |
1302 |
|
NM_001278543.1 |
NP_001265472.1
|
8, 9 |
1524 |
|
NM_001278544.1 |
NP_001265473.1
|
8, 9 |
1431 |
|
XM_017002235.1 |
XP_016857724.1
|
8, 9 |
1370 |
|
GenBank mRNA |
AK292740.1 |
- |
7, 8 |
1157 |
|
AK298425.1 |
- |
8, 9 |
1348 |
|
AK298444.1 |
- |
8, 9 |
1524 |
|
AK313382.1 |
- |
7, 8 |
1174 |
|
AK316209.1 |
- |
8, 9 |
1521 |
|
BC020701.1 |
- |
7, 8 |
1177 |
|
BC130575.1 |
- |
7, 8 |
1135 |
40146606 |
M16973.1 |
- |
7, 8 |
1137 |
|
X04393.1 |
- |
6, 7 |
972 |
|
View Full Product Details