Gene Transcripts

Gene Symbol
Atxn7l1
Entrez Gene ID
380753
Gene Name
ataxin 7-like 1
Gene Aliases
2810423G08Rik, Atxn7l4
Chromosome Location
Chr. 12: 33147639 - 33376408 on Build GRCm38
UniGene ID
Mm.352037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028139.5 NP_082415.2 4 1068
GenBank mRNA AK013145.1 - 4 975
BC017546.1 - 3 869

View Full Product Details