Gene Transcripts

Gene Symbol
Ccdc166
Entrez Gene ID
223648
Gene Name
coiled-coil domain containing 166
Gene Aliases
2410075B13Rik
Chromosome Location
Chr. 15: 75979872 - 75982285 on Build GRCm38
UniGene ID
Mm.17610
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163518.1 NP_001156990.1 1 1228
NM_146059.2 NP_666171.2 2 1115
GenBank mRNA AK019123.1 - 1 925
AK137409.1 - 1 929
AK171003.1 - 1 899
BC024536.1 - 1 836 4988004
BC047213.1 - 1 1229 6395374

View Full Product Details