Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195632.1 | NP_001182561.1 | 12 | 1169 | |
NM_177379.4 | NP_796353.3 | 3 | 505 | ||
GenBank mRNA | AB088417.1 | - | 4 | 452 | |
AY194286.1 | - | 3 | 232 | ||
BC132390.1 | - | 2 | 161 | 40130667 | |
BC138042.1 | - | 2 | 161 | 8861061 |