Gene Transcripts

Gene Symbol
Bpifc
Entrez Gene ID
270757
Gene Name
BPI fold containing family C
Gene Aliases
4732454E24, Bpil2
Chromosome Location
Chr. 10: 85959340 - 86015898 on Build GRCm38
UniGene ID
Mm.107214
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177772.4 NP_808440.2 3 249
GenBank mRNA AK028764.1 - 3 240
AK132518.1 - 3 285
BC112376.1 - 3 237 40057331
BY142313.1 - 3 249

View Full Product Details