Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004302.4 | NP_004293.1 | 7 | 1302 | |
NM_020327.3 | NP_064732.3 | 7 | 1254 | ||
NM_020328.3 | NP_064733.3 | 8 | 1425 | ||
XM_017020199.1 | XP_016875688.1 | 8 | 2123 | ||
XM_017020200.1 | XP_016875689.1 | 9 | 1518 | ||
GenBank mRNA | AK299120.1 | - | 8 | 1388 | |
AK299496.1 | - | 7 | 1254 | ||
BC000254.1 | - | 7 | 1251 | 3352720 | |
BC040531.1 | - | 7 | 1299 | 5260767 | |
BT007072.1 | - | 7 | 1225 | ||
U14722.1 | - | 7 | 1225 | ||
Z22536.1 | - | 7 | 1225 |