Gene Transcripts

Gene Symbol
ATXN1
Entrez Gene ID
6310
Gene Name
ataxin 1
Gene Aliases
ATX1, D6S504E, SCA1
Chromosome Location
Chr. 6: 16299112 - 16761490 on Build GRCh38
UniGene ID
Hs.434961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000332.3 NP_000323.2 9 2982
NM_001128164.1 NP_001121636.1 8 2933
GenBank mRNA BC113082.1 - 5 595
BC117125.1 - 2 2044 40125676
X79204.1 - 9 2949

View Full Product Details