Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206729.1 | NP_001193658.1 | 8 | 792 | |
NM_005465.4 | NP_005456.1 | 7 | 751 | ||
NM_181690.2 | NP_859029.1 | 7 | 751 | ||
XM_005272994.4 | XP_005273051.1 | 8 | 976 | ||
XM_005272995.2 | XP_005273052.1 | 8 | 782 | ||
XM_006711726.3 | XP_006711789.1 | 8 | 976 | ||
XM_011544012.2 | XP_011542314.1 | 8 | 976 | ||
XM_011544013.2 | XP_011542315.1 | 8 | 976 | ||
XM_016999985.1 | XP_016855474.1 | 9 | 924 | ||
GenBank mRNA | AF085234.1 | - | 7 | 686 | |
AF124141.1 | - | 7 | 651 | ||
AF135794.1 | - | 7 | 639 | ||
AJ245709.1 | - | 7 | 649 | ||
AK308052.1 | - | 7 | 677 | ||
AL117525.1 | - | 8 | 792 | ||
AY005799.1 | - | 7 | 712 | ||
BC020479.1 | - | 7 | 685 | ||
BC121154.2 | - | 7 | 722 | 40121374 |