Gene Transcripts

Gene Symbol
BTBD3
Entrez Gene ID
22903
Gene Name
BTB domain containing 3
Gene Aliases
dJ742J24.1
Chromosome Location
Chr. 20: 11890723 - 11926595 on Build GRCh38
UniGene ID
Hs.709366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282550.1 NP_001269479.1 5 1339
NM_001282551.1 NP_001269480.1 5 1435
NM_001282552.2 NP_001269481.1 4 1429
NM_001282554.1 NP_001269483.1 4 1344
NM_014962.3 NP_055777.1 4 1757
NM_181443.3 NP_852108.2 5 1520
XM_017027728.1 XP_016883217.1 6 1668
XM_017027729.1 XP_016883218.1 6 1600
XM_017027730.1 XP_016883219.1 3 1688
GenBank mRNA AB023169.1 - 4 1705
AK296364.1 - 3 983
AK299281.1 - 5 1520
AK300371.1 - 3 1040
AK307472.1 - 4 1087
AK316256.1 - 4 1320
AK316519.1 - 3 1263
AL137756.1 - 1 2509
BC093677.1 - 4 1228
BC101527.1 - 4 1227 8069033
BC109315.1 - 5 1368 40036410
BC109316.1 - 5 1368 40036411

View Full Product Details