Gene Transcripts

Gene Symbol
C6orf48
Entrez Gene ID
50854
Gene Name
chromosome 6 open reading frame 48
Gene Aliases
D6S57, G8
Chromosome Location
Chr. 6: 31834915 - 31839766 on Build GRCh38
UniGene ID
Hs.640836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040437.2 NP_001035527.1 1 216
NM_001040438.2 NP_001035528.1 1 216
NM_001287482.1 NP_001274411.1 1 216
NM_001287483.1 NP_001274412.1 1 216
NM_001287484.1 NP_001274413.1 1 216
GenBank mRNA AJ249731.1 - 1 225
AJ249732.1 - 1 225
AK311989.1 - 1 215
AL556045.3 - 1 182
BC000706.1 - 1 197 3349773
BC017660.1 - 2 114 4049404
BC066330.1 - 1 215
BG109213.1 - 1 144
BI114651.1 - 1 193
BJ996767.1 - 1 216
BQ920527.1 - 1 200
CD249104.1 - 1 231

View Full Product Details