Gene Transcripts
- Gene Symbol
- ASZ1
- Entrez Gene ID
- 136991
- Gene Name
- ankyrin repeat, SAM and basic leucine zipper domain containing 1
- Gene Aliases
- ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
- Chromosome Location
- Chr. 7: 117363222 - 117427523 on Build GRCh38
- UniGene ID
- Hs.352412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301821.1 |
NP_001288750.1
|
2, 3 |
279 |
|
NM_001301822.1 |
NP_001288751.1
|
2, 3 |
279 |
|
NM_130768.2 |
NP_570124.1
|
2, 3 |
279 |
|
GenBank mRNA |
AF461259.1 |
- |
2, 3 |
245 |
|
AK093445.1 |
- |
2, 3 |
232 |
|
BC028695.1 |
- |
2, 3 |
249 |
|
BC034963.1 |
- |
2, 3 |
254 |
4824818 |
BC126186.1 |
- |
2, 3 |
238 |
8991902 |
BC126188.1 |
- |
2, 3 |
238 |
8991904 |
BC144212.1 |
- |
2, 3 |
238 |
9052732 |
DB457846.1 |
- |
2, 3 |
279 |
|
View Full Product Details