Gene Transcripts
- Gene Symbol
- FAM219A
- Entrez Gene ID
- 203259
- Gene Name
- family with sequence similarity 219 member A
- Gene Aliases
- C9orf25
- Chromosome Location
- Chr. 9: 34398184 - 34458610 on Build GRCh38
- UniGene ID
- Hs.493771
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184940.1 |
NP_001171869.1
|
5, 6 |
708 |
|
NM_147202.1 |
NP_671735.1
|
5, 6 |
657 |
|
XM_005251402.4 |
XP_005251459.1
|
6, 7 |
915 |
|
XM_011517797.2 |
XP_011516099.1
|
5, 6 |
715 |
|
XM_011517798.2 |
XP_011516100.1
|
5, 6 |
664 |
|
XM_011517799.2 |
XP_011516101.1
|
5, 6 |
513 |
|
GenBank mRNA |
AK096350.1 |
- |
5, 6 |
657 |
|
AK123887.1 |
- |
6, 7 |
519 |
|
AK299882.1 |
- |
5, 6 |
703 |
|
BC025348.1 |
- |
1, 2 |
65 |
|
View Full Product Details