Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001197237.1 | NP_001184166.1 | 2 | 147 | |
NM_001197238.1 | NP_001184167.1 | 2 | 248 | ||
NM_001197239.1 | NP_001184168.1 | 2 | 125 | ||
NM_001197240.1 | NP_001184169.1 | 2 | 125 | ||
NM_006995.4 | NP_008926.2 | 2 | 125 | ||
NM_181531.2 | NP_853509.1 | 2 | 125 | ||
XM_005248797.3 | XP_005248854.1 | 2 | 142 | ||
XM_005248798.4 | XP_005248855.1 | 2 | 450 | ||
XM_006714953.3 | XP_006715016.1 | 2 | 394 | ||
XM_006714955.3 | XP_006715018.1 | 2 | 142 | ||
XM_011514228.2 | XP_011512530.1 | 2 | 447 | ||
XM_011514229.2 | XP_011512531.1 | 2 | 142 | ||
XM_011514231.2 | XP_011512533.1 | 2 | 450 | ||
XM_017010166.1 | XP_016865655.1 | 2 | 331 | ||
XM_017010167.1 | XP_016865656.1 | 2 | 395 | ||
XM_017010168.1 | XP_016865657.1 | 2 | 398 | ||
XM_017010169.1 | XP_016865658.1 | 2 | 116 | ||
GenBank mRNA | AK289883.1 | - | 2 | 113 | |
AK293526.1 | - | 2 | 100 | ||
AK298570.1 | - | 2 | 100 | ||
AK304744.1 | - | 2 | 124 | ||
BC013258.2 | - | 1 | 45 | ||
BC014021.2 | - | 2 | 123 | 4124138 | |
BC017497.1 | - | 2 | 140 | 4648935 | |
BI754184.1 | - | 2 | 100 | ||
DC373063.1 | - | 2 | 147 | ||
DC388932.1 | - | 2 | 126 | ||
U90550.1 | - | 2 | 100 |